Please login to your online account to display your discounted pricing
Fisherbrand™ Natural HDPE Wide-Mouth Round Bottles with White Polypropylene PE Foam Lined Caps
Suggested for use with water based chemicals and some food products. Fisherbrand™ Natural HDPE Wide-Mouth Round Bottles with White Polypropylene PE Foam Lined Cap feature a wide opening making them ideal for solids.
$25.40 - $1,011.03
Specifications
Liner Material PE Foam
Closure Material Polypropylene
Closure Type Screw
Closure Color White
Color Natural
View More Specs
Products
Catalog Number Mfr. No. Quantity Capacity (English) Capacity (Metric) Neck Finish Price Quantity    
02-911-990A
fisherbrand™
ALP263535
50/Cs. 68 oz. 2000mL 89-400 Case of 50 Each for $278.50
02-912-018A
fisherbrand™
ALP263546
180/Cs. 16 oz. 500mL 53-400 Case of 180 Each for $362.13
02-912-019A
fisherbrand™
ALP263550
850/Cs. 2 oz. 60mL 33-400 Case of 850 Each for $635.37
02-912-020A
fisherbrand™
ALP263552
72/Cs. 32 oz. 950mL 53-400 Case of 72 Each for $245.73
02-912-021A
fisherbrand™
ALP263532
475/Cs. 4 oz. 120mL 38-400 Case of 475 Each for $410.27
02-912-022A
fisherbrand™
ALP263537
364/Cs. 8.5 oz. 250mL 53-400 Case of 364 Each for $585.00
02-912-031A
fisherbrand™
ALP263549
48/Cs. 2 oz. 60mL 33-400 Case of 48 Each for $48.50
02-912-030A
fisherbrand™
ALP263547
24/Cs. 16 oz. 500mL 53-400 Case of 24 Each for $66.20
02-912-033A
fisherbrand™
ALP263536
6/Cs. 68 oz. 2000mL 89-400 Case of 6 Each for $40.23
02-912-034A
fisherbrand™
ALP263538
48/Cs. 8.5 oz. 250mL 53-400 Case of 48 Each for $95.27
02-912-032A
fisherbrand™
ALP263533
48/Cs. 4 oz. 120mL 38-400 Case of 48 Each for $56.00
02-912-036A
fisherbrand™
ALP263544
4/Cs. 128 oz. 3785mL 89-400 Case of 4 Each for $25.40
02-912-037A
fisherbrand™
ALP263543
48/Cs. 1 oz. 30mL 28-400 Case of 48 Each for $41.70
02-912-038A
fisherbrand™
ALP263551
12/Cs. 32 oz. 950mL 53-400 Case of 12 Each for $52.60
02-912-104A
fisherbrand™
ALP263542
1650/Cs. 1 oz. 30mL 28-400 Case of 1650 Each for $1,011.03
Description & Specifications
Specifications
Liner Material PE Foam
Closure Material Polypropylene
Closure Type Screw
Closure Color White
Color Natural
Material HDPE
Mouth Wide
Shape Round
  • Translucent bottles allow limited viewing of contents
  • Provides moisture barrier and are impact resistant