Please login to your online account to display your discounted pricing
Fisherbrand™ Natural HDPE Wide-Mouth Round Bottles without Cap
Ideal for general lab use with solids and powders. Fisherbrand™ Natural HDPE Wide Mouth Round Bottles without Cap features wide mouth that facilitates easy mixing, handling, and transfer of contents.
$25.33 - $420.70
Specifications
Color Natural
Material HDPE
Mouth Wide
Shape Round
View More Specs
Products
Catalog Number Mfr. No. Quantity Capacity (English) Capacity (Metric) Neck Finish Price Quantity    
02-911-874A
fisherbrand™
ALP263545
4/Cs. 128 oz. 3785mL 89-400 Case of 4 Each for $25.33
02-911-886A
fisherbrand™
ALP263534
475/Cs. 4 oz. 120mL 38-400 Case of 475 Each for $281.33
02-912-023A
fisherbrand™
ALP263553
72/Cs. 32 oz. 950mL 53-400 Case of 72 Each for $205.83
02-912-105A
fisherbrand™
ALP263539
364/Cs. 8.5 oz. 250mL 53-400 Case of 364 Each for $420.70
02-912-110A
fisherbrand™
ALP263548
180/Cs. 16 oz. 500mL 53-400 Case of 180 Each for $277.60
Description & Specifications
Specifications
Color Natural
Material HDPE
Mouth Wide
Shape Round
  • HDPE (high-density polyethylene) bottles provides moisture barrier, are translucent and impact resistant
  • For high-speed filling, demanding pharmaceutical specifications and rough handling of interstate shipping
No caps