Fisherbrand™ Natural HDPE Wide Mouth Round Bottles without Cap
Ideal for general lab use with solids and powders. Fisherbrand™ Natural HDPE Wide Mouth Round Bottles without Cap features wide mouth that facilitates easy mixing, handling, and transfer of contents.
Manufacturer:  Fisherbrand™ ALP263553
 View more versions of this product
Catalog No. 02-912-023A
Description
Description
Natural HDPE Wide Mouth Round Bottles are ideal for general lab use with solids and powders. These HDPE (high-density polyethylene) bottles provide an excellent moisture barrier, are translucent, and are impact resistant. These bottles are ideal for high-speed filling, demanding pharmaceutical specifications and the rough handling of interstate shipping. Available in tall style in 32oz size. Standard style available in a wide range of sizes, from 1oz to 128oz.
  • HDPE (high-density polyethylene) bottles provides moisture barrier, are translucent and impact resistant
  • For high-speed filling, demanding pharmaceutical specifications and rough handling of interstate shipping
Order Info
No caps
Specifications
Specifications
950mL
Natural, Natural
72/Cs.
Wide
Round
32 oz.
HDPE, HDPE
Bottle, Wide Mouth Round, Wide Mouth Round Bottle
53-400
Documents