Please login to your online account to display your discounted pricing
Fisherbrand™ Natural HDPE Wide-Mouth Round Bottles without Cap
Ideal for general lab use with solids and powders. Fisherbrand™ Natural HDPE Wide Mouth Round Bottles without Cap features wide mouth that facilitates easy mixing, handling, and transfer of contents.
Manufacturer: fisherbrand™  ALP263539
 View more versions of this product
Catalog No. 02-912-105A
  • / Case of 364 Each
Description & Specifications
Specifications
Type Bottle, Wide Mouth Round
Mouth Wide
Neck Finish 53-400
Shape Round
Quantity 364/Cs.
Capacity (Metric) 250mL
Material HDPE
Color Natural
Capacity (English) 8.5 oz.
  • HDPE (high-density polyethylene) bottles provides moisture barrier, are translucent and impact resistant
  • For high-speed filling, demanding pharmaceutical specifications and rough handling of interstate shipping