Please login to your online account to display your discounted pricing
Fisherbrand™ Natural HDPE Wide-Mouth Round Bottles without Cap
Ideal for general lab use with solids and powders. Fisherbrand™ Natural HDPE Wide Mouth Round Bottles without Cap features wide mouth that facilitates easy mixing, handling, and transfer of contents.
Manufacturer: fisherbrand™  ALP263534
 View more versions of this product
Catalog No. 02-911-886A
  • / Case of 475 Each
Description & Specifications
Specifications
Capacity (Metric) 120mL
Capacity (English) 4 oz.
Color Natural
Material HDPE
Quantity 475/Cs.
Type Bottle, Wide Mouth Round
Mouth Wide
Neck Finish 38-400
Shape Round
  • HDPE (high-density polyethylene) bottles provides moisture barrier, are translucent and impact resistant
  • For high-speed filling, demanding pharmaceutical specifications and rough handling of interstate shipping