Please login to your online account to display your discounted pricing
Fisherbrand™ Natural HDPE Wide-Mouth Oblong Bottles with White Polypropylene PE Foam Lined Cap
Ideal for sampling solids and powders. Fisherbrand™ Natural HDPE Wide-Mouth Oblong Bottles with White Polypropylene PE Foam Lined Cap offer a space saving design for the workbench.
$57.07 - $508.77
Specifications
Liner Material PE Foam
Closure Material Polypropylene
Closure Type Screw
Closure Color White
Color Natural
View More Specs
Includes:  White Polypropylene PE foam lined caps
Products
Catalog Number Mfr. No. Quantity Capacity (English) Capacity (Metric) Neck Finish Price Quantity    
02-911-948A
fisherbrand™
ALP263502
116/Cs. 32 oz. 1000mL 53-400 Case of 116 Each for $439.33
02-911-949A
fisherbrand™
ALP263503
12/Cs. 32 oz. 1000mL 53-400 Case of 12 Each for $57.07
02-911-952A
fisherbrand™
ALP263506
48/Cs. 4.2 oz. 125mL 38-400 Case of 48 Each for $63.37
02-911-953A
fisherbrand™
ALP263507
500/Cs. 4.2 oz. 125mL 38-400 Case of 500 Each for $508.77
02-911-954A
fisherbrand™
ALP263508
48/Cs. 5.5 oz. 160mL 38-400 Case of 48 Each for $63.80
02-911-956A
fisherbrand™
ALP263510
250/Cs. 8.5 oz. 250mL 43-400 Case of 250 Each for $381.23
02-911-959A
fisherbrand™
ALP263512
160/Cs. 16 oz. 500mL 43-400 Case of 160 Each for $304.30
02-911-960A
fisherbrand™
ALP263513
24/Cs. 16 oz. 500mL 43-400 Case of 24 Each for $63.33
02-911-957A
fisherbrand™
ALP263511
48/Cs. 8.5 oz. 250mL 43-400 Case of 48 Each for $91.33
Description & Specifications
Specifications
Liner Material PE Foam
Closure Material Polypropylene
Closure Type Screw
Closure Color White
Color Natural
Material HDPE
Mouth Wide
Shape Oblong
  • HDPE provides moisture barrier and are impact resistant
  • Ideal for high-speed filling, demanding pharmaceutical specifications and the rough handling of interstate shipping
  • Polypropylene closures with foamed polyethylene liners offer good chemical resistance including acids, alcohols, alkalis, aqueous products, cosmetics, household oils and solvents