Please login to your online account to display your discounted pricing
Fisherbrand™ Natural HDPE Wide Mouth Oblong Bottles without Cap
Ideal for sampling solids and powders. Fisherbrand™ Natural HDPE Wide Mouth Oblong Bottles without Cap offer a space saving design for the workbench.
Manufacturer: fisherbrand™  ALP263504
 View more versions of this product
Catalog No. 02-912-108A
  • / Case of 116 Each
Description & Specifications
Specifications
Capacity (Metric) 1000mL
Capacity (English) 32 oz.
Product Type Oblong Bottle
Color Natural
Material HDPE
Mouth Wide
Neck Finish 53-400
Quantity 116/Cs.
Shape Oblong
  • Features a wide mouth opening
  • HDPE material provides moisture barrier and are impact resistant
  • Ideal for high-speed filling, demanding pharmaceutical specifications and the rough handling of interstate shipping