Fisherbrand™ Natural HDPE Wide Mouth Oblong Bottles without Cap
Ideal for sampling solids and powders. Fisherbrand™ Natural HDPE Wide Mouth Oblong Bottles without Cap offer a space saving design for the workbench.
$239.67 - $383.10
Specifications
Color Natural
Material HDPE
Mouth Wide
Shape Oblong
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Neck Finish Quantity Capacity (English) Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Neck Finish Quantity Capacity (English) Capacity (Metric) Price Quantity  
02-912-106A Fisherbrand™
ALP263509
38-400 400/Cs. 5.5 oz. 160mL Case of 400 Each for $354.63
02-912-107A Fisherbrand™
ALP263514
43-400 160/Cs. 16 oz. 500mL Case of 160 Each for $239.67
02-912-108A Fisherbrand™
ALP263504
53-400 116/Cs. 32 oz. 1000mL Case of 116 Each for $383.10
Specifications
Description
  • Features a wide mouth opening
  • HDPE material provides moisture barrier and are impact resistant
  • Ideal for high-speed filling, demanding pharmaceutical specifications and the rough handling of interstate shipping
Order Info
Caps not included
Description & Specifications
Specifications
Natural
HDPE
Wide
Oblong