Fisherbrand™ Natural HDPE Narrow-Mouth Cylinder Bottles with White Polypropylene PE Foam Lined Cap
Ideal for handling liquid samples in the lab. Fisherbrand™ Natural HDPE Narrow-Mouth Cylinder Bottles with Polypropylene PE Foam Lined Cap are translucent and impact resistant.
$35.27 - $565.23
Specifications
Liner Material PE Foam
Closure Material Polypropylene
Closure Type Screw
Closure Color White
Color Natural
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Neck Finish Quantity Capacity (English) Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Neck Finish Quantity Capacity (English) Capacity (Metric) Price Quantity  
02-911-962A Fisherbrand™
ALP263455
24-410 336/Cs. 8 oz. 240mL Case of 336 Each for $349.03
02-911-979A Fisherbrand™
ALP263449
24-410 48/Cs. 2 oz. 60mL Case of 48 Each for $42.70
02-911-980A Fisherbrand™
ALP263454
24-410 48/Cs. 4 oz. 120mL Case of 48 Each for $42.70
02-911-981A Fisherbrand™
ALP263457
24-410 48/Cs. 8 oz. 240mL Case of 48 Each for $67.83
02-911-982A Fisherbrand™
ALP263446
24-410 24/Cs. 16 oz. 480mL Case of 24 Each for $50.40
02-911-983A Fisherbrand™
ALP263450
28-400 12/Cs. 32 oz. 960mL Case of 12 Each for $35.27
02-912-112A Fisherbrand™
ALP263448
24-410 900/Cs. 2 oz. 60mL Case of 900 Each for $565.23
02-912-293A Fisherbrand™
ALP263445
24-410 153/Cs. 16 oz. 480mL Case of 153 Each for $209.60
02-912-294A Fisherbrand™
ALP263451
28-400 66/Cs. 32 oz. 960mL Case of 66 Each for $141.97
Specifications
Description
Natural HDPE Narrow-Mouth Cylinder Bottles
  • Shoulder style allows to pour small quantities without spilling
  • Ideal for high-speed filling, demanding pharmaceutical specifications and the rough handling of interstate shipping
Polypropylene PE Foam/PTFE Lined Caps
  • Offers good impact strength, durability, thermal resistance, and pliability
  • Autoclavable
  • Liners consist of 3 plys: a low density foam core tightly sandwiched between two layers of polyethylene (LDPE)
  • Liners are very resilient and have strong resistance to chemicals and moisture and help keep bottle or jar contents free of odor and taste interference
Description & Specifications
Specifications
PE Foam
Screw
Natural
Narrow
Polypropylene
White
HDPE
Cylindrical