Please login to your online account to display your discounted pricing

Fisherbrand™ Natural HDPE Narrow-Mouth Cylinder Bottles with White Polypropylene PE Foam Lined Cap


Ideal for handling liquid samples in the lab. Fisherbrand™ Natural HDPE Narrow-Mouth Cylinder Bottles with Polypropylene PE Foam Lined Cap are translucent and impact resistant.

Manufacturer: fisherbrand™  ALP263451

 View more versions of this product

Polypropylene PE Foam lined cap

Catalog No. 02-912-294A

  • / Case of 66 Each
 

Description & Specifications

Specifications

Capacity (Metric) 960mL
Capacity (English) 32 oz.
Closure Material Polypropylene
Color Natural
Material HDPE
Quantity 66/Cs.
Type Bottle, Cylinder
Closure Color White
Liner Material PE Foam
Mouth Narrow
Neck Finish 28-400
Shape Cylindrical
Closure Type Screw

Natural HDPE Narrow-Mouth Cylinder Bottles

  • Shoulder style allows to pour small quantities without spilling
  • Ideal for high-speed filling, demanding pharmaceutical specifications and the rough handling of interstate shipping

Polypropylene PE Foam/PTFE Lined Caps

  • Offers good impact strength, durability, thermal resistance, and pliability
  • Autoclavable
  • Liners consist of 3 plys: a low density foam core tightly sandwiched between two layers of polyethylene (LDPE)
  • Liners are very resilient and have strong resistance to chemicals and moisture and help keep bottle or jar contents free of odor and taste interference