Please login to your online account to display your discounted pricing

Fisherbrand™ Natural HDPE Narrow-Mouth Cylinder Bottles with White Polypropylene PE Foam Lined Cap


Ideal for handling liquid samples in the lab. Fisherbrand™ Natural HDPE Narrow-Mouth Cylinder Bottles with Polypropylene PE Foam Lined Cap are translucent and impact resistant.

Manufacturer: fisherbrand™  ALP263448

 View more versions of this product

Polypropylene PE Foam lined cap

Catalog No. 02-912-112A

  • / Case of 900 Each
 

Description & Specifications

Specifications

Capacity (Metric) 60mL
Capacity (English) 2 oz.
Product Type Cylinder Bottle
Closure Color White
Closure Material Polypropylene
Closure Type Screw
Color Natural
Material HDPE
Liner Material PE Foam
Mouth Narrow
Neck Finish 24-410
Quantity 900/Cs.
Shape Cylindrical

Natural HDPE Narrow-Mouth Cylinder Bottles

  • Shoulder style allows to pour small quantities without spilling
  • Ideal for high-speed filling, demanding pharmaceutical specifications and the rough handling of interstate shipping
Polypropylene PE Foam/PTFE Lined Caps
  • Offers good impact strength, durability, thermal resistance, and pliability
  • Autoclavable
  • Liners consist of 3 plys: a low density foam core tightly sandwiched between two layers of polyethylene (LDPE)
  • Liners are very resilient and have strong resistance to chemicals and moisture and help keep bottle or jar contents free of odor and taste interference