Please login to your online account to display your discounted pricing

Fisherbrand™ Natural HDPE Narrow-Mouth Cylinder Bottles without Cap

Ideal for pharmaceutical applications. Fisherbrand™ Natural HDPE Narrow-Mouth Cylinder Bottles without Cap are perfect for handling liquid samples in the lab.

$116.90 - $249.20

Specifications

Color Natural
Material HDPE
Mouth Narrow
Shape Cylindrical
Closure Included No
View More Specs
Products
Catalog Number Mfr. No. Quantity Capacity (English) Capacity (Metric) Neck Finish Price Quantity    

02-911-800A

 
fisherbrand™
ALP263447
153/Cs. 16 oz. 480mL 24-410 Case of 153 Each for $151.33

02-911-963A

 
fisherbrand™
ALP263456
336/Cs. 8 oz. 240mL 24-410 Case of 336 Each for $226.73

02-911-988A

 
fisherbrand™
ALP263452
66/Cs. 32 oz. 960mL 28-400 Case of 66 Each for $116.90

02-911-989A

 
fisherbrand™
ALP263453
500/Cs. 4 oz. 120mL 24-410 Case of 500 Each for $249.20
Description & Specifications

Specifications

Color Natural
Material HDPE
Mouth Narrow
Shape Cylindrical
Closure Included No
For Use With (Application) Handling liquid samples in the lab

  • Shoulder style allows to pour small quantities without spilling
  • HDPE bottles are ideal for high-speed filling, demanding pharmaceutical specifications and rough handling of interstate shipping

Caps not included