Please login to your online account to display your discounted pricing

Fisherbrand™ Natural HDPE Narrow-Mouth Cylinder Bottles without Cap


Ideal for pharmaceutical applications. Fisherbrand™ Natural HDPE Narrow-Mouth Cylinder Bottles without Cap are perfect for handling liquid samples in the lab.

Manufacturer: fisherbrand™  ALP263456

 View more versions of this product

Catalog No. 02-911-963A

  • / Case of 336 Each
 

Description & Specifications

Specifications

Capacity (Metric) 240mL
Capacity (English) 8 oz.
Closure Included No
Product Type Cylinder Bottle
Color Natural
For Use With (Application) Handling liquid samples in the lab
Material HDPE
Mouth Narrow
Neck Finish 24-410
Bottle Material HDPE
Quantity 336/Cs.
Shape Cylindrical

  • Shoulder style allows to pour small quantities without spilling
  • HDPE bottles are ideal for high-speed filling, demanding pharmaceutical specifications and rough handling of interstate shipping