Please login to your online account to display your discounted pricing

Fisherbrand™ Natural HDPE Narrow-Mouth Cylinder Bottles without Cap


Ideal for pharmaceutical applications. Fisherbrand™ Natural HDPE Narrow-Mouth Cylinder Bottles without Cap are perfect for handling liquid samples in the lab.

Manufacturer: fisherbrand™  ALP263447

 View more versions of this product

Catalog No. 02-911-800A

  • / Case of 153 Each
 

Description & Specifications

Specifications

Capacity (Metric) 480mL
Capacity (English) 16 oz.
Closure Included No
Product Type Cylinder Bottle
Color Natural
For Use With (Application) Handling liquid samples in the lab
Material HDPE
Mouth Narrow
Neck Finish 24-410
Bottle Material HDPE
Quantity 153/Cs.
Shape Cylindrical

  • Shoulder style allows to pour small quantities without spilling
  • HDPE bottles are ideal for high-speed filling, demanding pharmaceutical specifications and rough handling of interstate shipping