Thermo Scientific™ National Vial Crushers
Reduce waste disposal costs by reducing the total volume of waste with the Thermo Scientific™ National Vial Crusher.
Manufacturer:  Thermo Scientific™ C4040PVCM
 View more versions of this product
Catalog No. 03-396-93
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Description
Reduce waste disposal costs by reducing the total volume of waste with the Thermo Scientific™ National Vial Crusher.
  • Breaks and separates liquid from all 1mL to 100mL vials and ampules
  • Capable of processing approximately 5000 2mL vials per hour
  • Safe use in a fume hood or connect to a vacuum for limited exposure to flammable or toxic vapors
  • Environmentally responsible waste handling—crushed glass is automatically separated from solvent for separate disposal
Description & Specifications
Specifications
Pneumatic Benchtop Vial Crusher with Stainless-steel Mobile Cart
Vial Accessories
Pneumatic benchtop vial crusher with stainless-steel mobile cart