Please login to your online account to display your discounted pricing
Thermo Scientific™ National Unassembled Target LoVial™ Wide-Opening Vial Kits
Keep all vials and compatible crimp seals handy and dust-free with Thermo Scientific™ National Unassembled Target LoVial Wide-Opening Vial Kits.
Manufacturer: thermo scientific™  C401188AW
 View more versions of this product
Catalog No. 03-375-4D
  • / Pack of 500 Each
Description & Specifications
Specifications
Type Unassembled Vial Kit
Vial Volume (Metric) 4mL
Septa PTFE/Red Rubber
Color Amber ID
Item Description Amber I-D; PTFE/Red Rubber
Closure Material Aluminum
Closure Color Silver
Description Amber Glass; ID patchSilver Seal, PTFE/Red Rubber Septum; 4mL
Keep all vials and compatible crimp seals handy and dust-free with Thermo Scientific™ National Unassembled Target LoVial Wide-Opening Vial Kits.
  • Two-compartment benchtop storage tray
  • Include matched quantities of vials and silver aluminum seals with prefitted septa