Please login to your online account to display your discounted pricing
Thermo Scientific™ National Unassembled Headspace Convenience Kits
Thermo Scientific™ National Unassembled Headspace Convenience Kits are available for the most popular vial and seal combinations.
$228.26 - $228.26
Specifications
Description 20mL round bottom vials; Septa: PTFE/Butyl, Pressure Release
Type Unassembled Vial Kit
Item Description C4020-143
Septa PTFE/Butyl, Pressure Release
Vial Volume (Metric) 20mL
View More Specs
Products
Catalog Number Mfr. No. Septa Price Quantity    
03-397-42
thermo scientific™
C4020143
PTFE/Butyl, Pressure Release Case of 100 Each for $228.26
Description & Specifications
Specifications
Description 20mL round bottom vials; Septa: PTFE/Butyl, Pressure Release
Type Unassembled Vial Kit
Item Description C4020-143
Septa PTFE/Butyl, Pressure Release
Vial Volume (Metric) 20mL
Thermo Scientific™ National Unassembled Headspace Convenience Kits are available for the most popular vial and seal combinations.
  • 20mL round bottom vials
Includes:
100 vials and 100 seals with prefitted septa