Please login to your online account to display your discounted pricing
Thermo Scientific™ National Unassembled Certified™ Vial Kits
Thermo Scientific™ National Unassembled Certified Vial Kits provide extra assurance that time after time, the product upholds rigorous manufacturing standards.
Manufacturer: thermo scientific™  CERT401586
 View more versions of this product
Catalog No. 03-377-354
  • / Pack of 100 Each
Description & Specifications
Specifications
Closure Material Bonded PTFE/Silicone
Item Description 4mL Amber Screw-top Vial Kit
Septa PTFE/Silicone
Description Certified 4mL Amber Glass Screw-top Vial Kit, 13-425 Screw Thread
Color Amber
Closure Type Screw Thread
Type Unassembled Vial Kit
Vial Closure Size 13-425mm
Vial Volume (Metric) 4mL
Thermo Scientific™ National Unassembled Certified Vial Kits provide extra assurance that time after time, the product upholds rigorous manufacturing standards.
  • Fully lot tested for HPLC and GC analysis
  • Certificate of Conformance in every pack
  • Bonded caps to prevent septa push-through
  • Clear Type 1, 33 expansion borosilicate glass or amber Type 2, 51 expansion borosilicate glass
  • Available with or without ID patch
  • Fixed micro vials or inserts for limited-volume sampling