Please login to your online account to display your discounted pricing

Thermo Scientific National Unassembled Certified™ Vial Kits


Thermo Scientific™ National Unassembled Certified Vial Kits provide extra assurance that time after time, the product upholds rigorous manufacturing standards.

Manufacturer: thermo scientific  CERT400080

 View more versions of this product

Catalog No. 03-377-345

  • / Pack of 100 Each
 

Description & Specifications

Specifications

Septa PTFE/Rubber
Color Clear
Type Unassembled Vial Kit
Vial Closure Size 9-425mm
Vial Volume (Metric) 2mL
Closure Material PTFE/Rubber
Closure Type Screw Thread
Item Description DP 2mL Clear; PTFE/Rubber
Description Certified Target DP 2mL Clear Vial, 9mm Screw Thread

Thermo Scientific™ National Unassembled Certified Vial Kits provide extra assurance that time after time, the product upholds rigorous manufacturing standards.

  • Fully lot tested for HPLC and GC analysis
  • Certificate of Conformance in every pack
  • Bonded caps to prevent septa push-through
  • Clear Type 1, 33 expansion borosilicate glass or amber Type 2, 51 expansion borosilicate glass
  • Available with or without ID patch
  • Fixed micro vials or inserts for limited-volume sampling