Please login to your online account to display your discounted pricing
Thermo Scientific National Target™ Unassembled Vial, Cap and Septum Convenience Kits
Keep vials, caps and septa within easy reach and dust free with Thermo Scientific™ National Target Unassembled Convenience Kits.
Manufacturer: thermo scientific  C4010S92
 View more versions of this product
Catalog No. 03-376-402
  • / Pack of 100 Each
Description & Specifications
Specifications
Color Clear Silanized
Pre-slit Pre-slit
Type Unassembled Vial Kit
Item Description Clear Silanized; PTFE/Silicone—Pre-slit
Vial Volume (Metric) 2mL
Closure Color Light Blue
Septa PTFE/Silicone - Pre-slit
Description Clear Glass; Silanized; Light Blue Cap, Pre-Slit Blue PTFE/White SiliconeSeptum; 2mL
Keep vials, caps and septa within easy reach and dust free with Thermo Scientific™ National Target Unassembled Convenience Kits.
  • Two-compartment benchtop storage tray
  • I-D™ vials feature write-on patch with graduations
  • All items remain handy and dust-free
  • Caps with septa are not attached to vials (packed in separate polybag)
Includes:
100 vials and 100 caps with pre-assembled septa