Please login to your online account to display your discounted pricing
Thermo Scientific™ National Target™ Unassembled Vial, Cap and Septum Convenience Kits
Keep vials, caps and septa within easy reach and dust free with Thermo Scientific™ National Target Unassembled Convenience Kits.
Manufacturer: thermo scientific™  C401088WBLK
 View more versions of this product
Catalog No. 03-339-355
  • / Pack of 100 Each
Description & Specifications
Specifications
Septa PTFE/Silicone
Color Clear ID
Type Unassembled Vial Kit
Closure Color Black
Item Description Clear ID; PTFE/Silicone
Description Clear Glass I-D; Black cap; PTFE/Silicone septum
Keep vials, caps and septa within easy reach and dust free with Thermo Scientific™ National Target Unassembled Convenience Kits.
  • Two-compartment benchtop storage tray
  • I-D™ vials feature write-on patch with graduations
  • All items remain handy and dust-free
  • Caps with septa are not attached to vials (packed in separate polybag)
Includes:
100 vials and 100 caps with pre-assembled septa