Please login to your online account to display your discounted pricing
Thermo Scientific™ National Target™ Snap-It™ Kits
Thermo Scientific™ National Target Snap-It Convenience Kits provide vials and Snap-it caps with prefitted septa in storage trays.
Manufacturer: thermo scientific™  C401174
 View more versions of this product
Catalog No. 03-377-88
  • / Pack of 100 Each
Description & Specifications
Specifications
Septa PTFE/Silicone, Pre-slit
Pre-slit Pre-slit
Color Clear
Type Assembled Vial Kit
Vial Volume (Metric) 2mL
Closure Color Clear
Closure Material PTFE/Silicone, Pre-slit
Item Description Clear Glass; PTFE/Silicone, Pre-Slit
Description Clear Glass; Septum: PTFE/Silicone, Pre-Slit
Thermo Scientific™ National Target Snap-It Convenience Kits provide vials and Snap-it caps with prefitted septa in storage trays.
  • 2mL,12 × 32mm vials
  • Choice of vial and preassembled seals with septa
  • Packaged in two compartment benchtop storage trays
  • Include matched quantities of vials and Snap-It caps with prefitted septa
  • All items remain dust-free