missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ National Target™ Empty Caps without Septum for 10-425 Vials

Thermo Scientific™ National Target Empty Open-Top Caps for 10-425 Vials are designed to be used with any of our 10mm septa.

$17.49 - $17.58

Specifications

Closure Material Polypropylene
Type Open Top Screw Cap
Vial Closure Size 10-425mm
For Use With 10-425 Vials
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Closure Color Price Quantity  
Catalog Number Mfr. No. Closure Color Price Quantity  
03-376-446 Thermo Scientific
C401098G
Green Pack of 100 Each for $17.49
03-376-473 Thermo Scientific
C401098Y
Yellow Pack of 100 Each for $17.58
Specifications

Description

Thermo Scientific™ National Target Empty Open-Top Caps for 10-425 Vials are designed to be used with any of our 10mm septa.

  • Polypropylene caps are chemically inert and suitable for most chromatography applications
  • Target 10-425 caps feature a flanged rim, making them suitable for Japanese autosampler trays
Description & Specifications

Specifications

Polypropylene
Open Top Screw Cap
10-425mm
10-425 Vials