missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ National Target DP™ Screw Thread Caps

Thermo Scientific™ National Target DP Screw Thread Caps with open top are designed for use with Target DP septa.

$13.25 - $17.62

Specifications

Closure Material Polypropylene
Septum No Septum
Type Open Top Screw Cap
For Use With Target DP septa
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Closure Color Septum Price Quantity  
Catalog Number Mfr. No. Closure Color Septum Price Quantity  
03-377-4B Thermo Scientific™
C500098G
Green No Septum Pack of 100 Each for $17.62
03-377-4C Thermo Scientific™
C500098R
Red No Septum Pack of 100 Each for $13.25
Case of 10 Pack for $132.50
03-377-4D Thermo Scientific™
C500098Y
Yellow No Septum Pack of 100 Each for $13.25
Case of 10 Pack for $132.50
Specifications

Description

Thermo Scientific™ National Target DP Screw Thread Caps with open top are designed for use with Target DP septa.

  • Polypropylene, open-top caps for use with Target DP septa
Description & Specifications

Specifications

Polypropylene
No Septum
Open Top Screw Cap
Target DP septa