Please login to your online account to display your discounted pricing
Thermo Scientific™ National Target™ Assembled Vial, Cap and Septum Kits
Thermo Scientific™ National Target Assembled Vial Kits include vials with pre-attached caps and septa, all packaged in convenient trays or polybags.
Manufacturer: thermo scientific™  C401019AW
 View more versions of this product
Catalog No. 03-376-433
  • / Pack of 100 Each
Description & Specifications
Specifications
Septa PTFE/Silicone/PTFE
Color Amber ID
Type Assembled Vial Kit
Closure Color Light Blue
Item Description Amber ID; Blue Cap; PTFE/Silicone/PTFE
Description Amber Glass I-D; Light Blue Cap; PTFE/Silicone/PTFE septum
Thermo Scientific™ National Target Assembled Vial Kits include vials with pre-attached caps and septa, all packaged in convenient trays or polybags.
  • Assembled kits include 100 vials with pre-attached caps and septa
  • Packaged in convenient vial trays with clear covers or in polybags
  • Vial trays are stackable for easy storage
  • Vial trays hold vials upright for easy filling and maintaining sample order