Please login to your online account to display your discounted pricing
Thermo Scientific™ National Target™ Assembled Vial, Cap and Septum Kits
Thermo Scientific™ National Target Assembled Vial Kits include vials with pre-attached caps and septa, all packaged in convenient trays or polybags.
Manufacturer: thermo scientific™  C401021W
 View more versions of this product
Catalog No. 03-375-14L
  • / Pack of 100 Each
Description & Specifications
Specifications
Color Clear ID
Closure Color Light Blue
Item Description Clear ID; Blue Cap; White virgin PTFE
Description Clear Glass I-D; Light Blue Cap; White virgin PTFE septum
Septa White virgin PTFE
Type Assembled Vial Kit
Thermo Scientific™ National Target Assembled Vial Kits include vials with pre-attached caps and septa, all packaged in convenient trays or polybags.
  • Assembled kits include 100 vials with pre-attached caps and septa
  • Packaged in convenient vial trays with clear covers or in polybags
  • Vial trays are stackable for easy storage
  • Vial trays hold vials upright for easy filling and maintaining sample order