Please login to your online account to display your discounted pricing
Thermo Scientific™ National Target™ 10mm Septa for 10-425 Threaded Caps
Thermo Scientific™ National Target 10mm Septa are designed for use with 10-425 threaded caps.
Manufacturer: thermo scientific™  C401065
 View more versions of this product
Catalog No. 03-397-06
  • / Pack of 100 Each
Description & Specifications
Specifications
Material Red PTFE/White Silicone, Star-slit
Vial Closure Size 10-425mm
Type Screw Cap Septa
For Use With Vial Screw Caps
Vial Closure Size 10-425mm
Thermo Scientific™ National Target 10mm Septa are designed for use with 10-425 threaded caps.