Thermo Scientific™ National Snap-It™ Closures for 12 × 32 crimp/snap vials
Thermo Scientific™ National Snap-It Closures allow quick, easy application and removal from Target Snap-It vials.
$13.89 - $78.23
Specifications
Type Snap Cap
Vial Closure Size 11mm
Vial Volume (Metric) 2mL
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Closure Color Septum Price Quantity  
Catalog Number Mfr. No. Closure Color Septum Price Quantity  
03-375-24J Thermo Scientific™
C401150Y
Yellow Integral PE Pack of 100 Each for $13.89
Case of 10 Pack for $138.90
03-396N Thermo Scientific™
C401153G
Green Red PTFE/White Silicone/Red PTFE Pack of 100 Each for $78.23
Case of 10 Pack for $782.30
Specifications
Description
Thermo Scientific™ National Snap-It Closures allow quick, easy application and removal from Target Snap-It vials.
  • Caps eliminate the need for crimping or de-capping tools
  • Easy to apply and easy to remove from vials
  • Available in a variety of colors and septa choices
Description & Specifications
Specifications
Snap Cap
11mm
2mL