Thermo Scientific™ National Snap-It™ Closures for 12 × 32 crimp/snap vials
Thermo Scientific™ National Snap-It Closures allow quick, easy application and removal from Target Snap-It vials.
Manufacturer:  Thermo Scientific™ C401153G
 View more versions of this product
Catalog No. 03-396N
Specifications
Description
Thermo Scientific™ National Snap-It Closures allow quick, easy application and removal from Target Snap-It vials.
  • Caps eliminate the need for crimping or de-capping tools
  • Easy to apply and easy to remove from vials
  • Available in a variety of colors and septa choices
Description & Specifications
Specifications
2mL
11mm
Green
C4011-53G
Snap Cap
Red PTFE/White Silicone/Red PTFE