Please login to your online account to display your discounted pricing

Thermo Scientific™ National Snap-It™ Closures for 12 × 32 crimp/snap vials


Thermo Scientific™ National Snap-It Closures allow quick, easy application and removal from Target Snap-It vials.

Manufacturer: thermo scientific™  C401150Y

 View more versions of this product

Catalog No. 03-375-24J

  • / Pack of 100 Each
  • / Case of 10 Pack
 

Description & Specifications

Specifications

Item Description C4011-50Y
Closure Color Yellow
Septum Integral PE
Vial Closure Size 11mm
Type Snap Cap
Vial Volume (Metric) 2mL

Thermo Scientific™ National Snap-It Closures allow quick, easy application and removal from Target Snap-It vials.

  • Caps eliminate the need for crimping or de-capping tools
  • Easy to apply and easy to remove from vials
  • Available in a variety of colors and septa choices