Thermo Scientific™ National Septa for 4mL Screw-Thread Vials
Thermo Scientific™ National Vial Septa are designed for use with 4mL screw-thread vials.
$79.58 - $79.58
Specifications
Material Blue PTFE/White Silicone—Pre-slit
Type Single slit septum
For Use With Vial Screw Caps
Item Description Blue PTFE/White Silicone; Preslit
Vial Closure Size 13-425mm
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Material Type Price Quantity  
Catalog Number Mfr. No. Material Type Price Quantity  
03-377-112 Thermo Scientific™
C401555
Blue PTFE/White Silicone—Pre-slit Single slit septum Pack of 100 Each for $79.58
Specifications
Description
Thermo Scientific™ National Vial Septa are designed for use with 4mL screw-thread vials.
  • PTFE disk septa are chemically inert
  • PFTE/red rubber septa are economical for many GC applications
  • PTFE/silicone septa are most popular for HPLC and IC applications
  • Pre-slit septa are easier to pierce with thin gauge needles
Description & Specifications
Specifications
Blue PTFE/White Silicone—Pre-slit
Vial Screw Caps
13-425mm
Single slit septum
Blue PTFE/White Silicone; Preslit