Thermo Scientific™ National SepCap™ Polyspring™ Limited Volume Inserts for Waters 96-Position Autosampler
Prevent the syringe from bottoming out and damaging the needle with Thermo Scientific™ National SepCap Polyspring Limited-Volume Inserts for Waters 96-Position Autosampler.
$154.48 - $154.48
Specifications
Type Polyspring Conical, Silanized
Vial Volume (Metric) 250µL
Features Polyspring Conical, Silanized
For Use With Waters 96-Position Autosampler
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Type Price Quantity  
Catalog Number Mfr. No. Type Price Quantity  
03-375-1BS Thermo Scientific™
C4015S96A
Polyspring Conical, Silanized Pack of 100 Each for $154.48
Specifications
Description
Prevent the syringe from bottoming out and damaging the needle with Thermo Scientific™ National SepCap Polyspring Limited-Volume Inserts for Waters 96-Position Autosampler.
  • Type I, borosilicate glass
Description & Specifications
Specifications
Polyspring Conical, Silanized
250µL
Polyspring Conical, Silanized
Waters 96-Position Autosampler