Please login to your online account to display your discounted pricing
Thermo Scientific™ National Scientific Polyethylene SepCap™ Cap
With built-in septa
$48.60 - $69.40
Specifications
Color Natural
Material Closure Polypropylene
View More Specs
Products
Catalog Number Mfr. No. Type Price Quantity    
03-391A
thermo fisher scientific
C401181
12 x 32 replacement Closure Pack of 500 Each for $69.40
03-375-2H
thermo scientific™
C401597
Closure For shell vials Pack of 500 Each for $48.60
Description & Specifications
Specifications
Color Natural
Material Closure Polypropylene
  • Natural color
  • Polyethylene with 0.010 in. built-in septa