Please login to your online account to display your discounted pricing

National Scientific BioStor™ Screw Cap Vials, Skirted


Thick-walled construction, with reinforced support collar, allows high speed centrifugation to 17,000xg

Manufacturer: national scientific  BS20NABP

 View more versions of this product

Catalog No. 11-844-18

  • / Case of 1000 Each
 

Description & Specifications

Specifications

Closure Type Screw cap
Type Skirted Vial
For Use With (Application) Collection, processing, centrifugation, long term storage and OEM packaging or shipping of restriction enzymes or other DNA or RNA modifying reagents
Includes Reinforced support collar, flexible ethylene or propylene O-ring, external rib, circular 3mm syringe port
Temperature Range (English) 249° to -319°F
Color Natural
Capacity 0.06 oz., 2mL
Temperature Range (Metric) 121° to -195°C
Capacity (English) 0.06 oz.
Quantity 1000/Cs.
Capacity (Metric) 2mL