missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
National Scientific™ BioStor™ Screw Cap Vials, Skirted
Thick-walled construction, with reinforced support collar, allows high speed centrifugation to 17,000xg
$338.13 - $338.13
Specifications
For Use With (Application) | Collection, processing, centrifugation, long term storage and OEM packaging or shipping of restriction enzymes or other DNA or RNA modifying reagents |
---|---|
Temperature Range (English) | -319°F to +249°F |
Temperature Range (Metric) | -195°C to +121°C |
Closure Type | Screw Cap |
Catalog Number | Mfr. No. | Volume (Metric) | Color | Sterility | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Color | Sterility | Price | Quantity | |||
11-844-18
|
National Scientific™
BS20NABP |
2 mL | Natural | Non-sterile |
Case of 1000 Each for $338.13
|
|
Description
Multi-purpose design serves a broad spectrum of bio-technology laboratory requirements including the collection, processing, centrifugation, long-term storage, and OEM packaging/shipping of restriction enzymes or other DNA/RNA modifying reagents.
- Circular 3mm syringe port in cap which is easily pierced for radioisotope labeling and fraction collecting
- Inert, LN2-grade polypropylene protects critical sample viability at extreme temperatures ranging from +121C to -195C (LN2 vapor phase)
- Two-turn closure design ensures that tightening of unique screw-cap will uniformly compress “O”-ring against beveled seat, eliminating distortion and maintaining seal integrity
- Vials, caps and ethylene/propylene “O”-rings chemial-resistant and autoclavable with cap loosened
- Molded-in volume lines and etched writing panels on vials and caps will not crack off or dissolve
- External ribs on vials position and lock into serrated holes in metal BioStor Racks, allowing single-handed opening and closing of screw-caps
- Available in natural and rainbow pack
Packaging:
- Nonsterile: 500/bag, 2 bags/case
- Sterile: 50 assembled caps w/ vials per bag, 10 bags/case
Specifications
Collection, processing, centrifugation, long term storage and OEM packaging or shipping of restriction enzymes or other DNA or RNA modifying reagents | |
-319°F to +249°F | |
-195°C to +121°C | |
Screw Cap |