National Scientific™ BioStor™ Multi-Purpose Specimen Containers
These containers with re-sealable lids can be used to store almost any liquid, powder or solid, making them ideal for laboratory or hospital use.
Manufacturer:  National Scientific™ MPCET008
 View more versions of this product
Catalog No. 11-848-3
Description
Description
Injection molded of polypropylene or linear polyethylene, these containers are virtually indestructible in normal use
  • Inert to most lab chemicals including formaldehyde, weak acids, organic solvents and all bases
  • Stable, wide-mouth design allows for ease of filling and is less likely to tip over, even when stacked
  • Tight-fitting, snap-cap lids keep odors in and minimize chance of leakage
  • Contents are visible on contact with translucent plastic walls
  • Resistant to boiling and freezing temperatures
Specifications
Specifications
HDPE
8.0 oz.
Specimen container with snap-on lid
Tall
3 in.