Please login to your online account to display your discounted pricing
Thermo Scientific National Polypropylene and Glastic™ Vials with 8-425 Finish
Thermo Scientific™ National Polypropylene and Glastic Vials provide an economical alternative to glass for some applications.
Manufacturer: thermo scientific  C401315GST
 View more versions of this product
Catalog No. 03-378-340
  • / Pack of 100 Each
Description & Specifications
Specifications
Material TPX/Glass Insert
Vial Volume (Metric) 450µL
Volume (Metric) Working 300µL
Item Description Glastic Glass Insert/ TPX MicroVials
Vial Closure Size 8-425mm
Diameter 12mm
Residual Volume <2µL
Quantity 100/Pk.
Type Screw Thread Vial
Description 150µL Glastic Glass Insert/ TPX MicroVials
Color Clear
Thermo Scientific™ National Polypropylene and Glastic Vials provide an economical alternative to glass for some applications.
  • Precision formed tips are designed to aid sample recovery for limited-volume applications
  • Glastic vial features a glass insert pre-inserted inside a clear TPX vial