Please login to your online account to display your discounted pricing
Thermo Scientific™ National Plate +™ Glass-Coated Microplates
Thermo Scientific™ National Plate + Glass-Coated Microplates provide a cost-effective alternative to solid glass plates.
$1,108.09 - $1,152.83
Specifications
Material PP
Surface Coating 200nm thick layer of silicone dioxide
View More Specs
Products
Catalog Number Mfr. No. Volume (Metric) Well Price Quantity    
03-398-27
thermo scientific
P96F300G
300µL Pack of 10 Each for $1,108.09
03-398-26
thermo scientific™
P96U250G
250µL Pack of 10 Each for $1,152.83
Description & Specifications
Specifications
Material PP
Surface Coating 200nm thick layer of silicone dioxide
Thermo Scientific™ National Plate + Glass-Coated Microplates provide a cost-effective alternative to solid glass plates.
  • Lightweight, precision-molded, cost-effective alternative to solid glass plates
  • Polypropylene microplates coated with 200nm thick layer of silicone dioxide
  • Chemically resistant with the qualities of glass and advantages of polypropylene
  • Operating temperatures of -80°C to 80°C