Thermo Scientific™ National Open-Top Caps without Septum for 4mL Screw Thread Vials
Thermo Scientific™ National Open-Top Caps without Septum for 4mL Screw Thread Vials are designed with open-top for use with any of our 13mm septa.
$115.57 - $123.58
Specifications
Closure Material Polypropylene
Type Open-Top Cap
Vial Closure Size 13-425mm
For Use With 4mL Screw Thread Vials
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Closure Color Closure Material Price Quantity  
Catalog Number Mfr. No. Closure Color Closure Material Price Quantity  
03-378-384 Thermo Scientific™
C401597B
Blue Polypropylene Case of 1000 Each for $123.58
03-378-385 Thermo Scientific™
C401597G
Green Polypropylene Case of 1000 Each for $115.57
03-378-386 Thermo Scientific™
C401597R
Red Polypropylene Case of 1000 Each for $115.57
Specifications
Description
Thermo Scientific™ National Open-Top Caps without Septum for 4mL Screw Thread Vials are designed with open-top for use with any of our 13mm septa.
  • Open-top
  • 13-425 GPI Thread
Description & Specifications
Specifications
Polypropylene
Open-Top Cap
13-425mm
4mL Screw Thread Vials