Please login to your online account to display your discounted pricing
Thermo Scientific™ National Open-Top Caps without Septum for 4mL Screw Thread Vials
Thermo Scientific™ National Open-Top Caps without Septum for 4mL Screw Thread Vials are designed with open-top for use with any of our 13mm septa.
Manufacturer: thermo scientific™  C401597R
 View more versions of this product
Catalog No. 03-378-386
  • / Case of 1000 Each
Description & Specifications
Specifications
Vial Closure Size 13-425mm
Closure Color Red
Closure Material Polypropylene
For Use With 4mL Screw Thread Vials
Description Red; Polypropylene
Type Open-Top Cap
Item Description Red; Polypropylene
Thermo Scientific™ National Open-Top Caps without Septum for 4mL Screw Thread Vials are designed with open-top for use with any of our 13mm septa.
  • Open-top
  • 13-425 GPI Thread