Please login to your online account to display your discounted pricing

Thermo Scientific™ National Inserts for Target LoVial™ Wide-Opening Vials

With a nominal O.D. of 6mm, these Thermo Scientific™ National Inserts fit Target LoVial Wide-Opening Vials.

$1,228.89 - $1,228.89

Specifications

Type Insert is suspended from the rim of the vial
Vial Volume (Metric) 150µL
Features Insert is suspended from the rim of the vial
For Use With Target™ LoVial™ Wide-Opening Vials
Item Description 150uL Glass Conical with Top Spring
View More Specs
Products
Catalog Number Mfr. No. Type Vial Volume (Metric) Price Quantity    

03-378-383

 
thermo scientific™
C4010630TS
Insert is suspended from the rim of the vial 150µL Pack of 500 Each for $1,228.89
Description & Specifications

Specifications

Type Insert is suspended from the rim of the vial
Vial Volume (Metric) 150µL
Features Insert is suspended from the rim of the vial
For Use With Target™ LoVial™ Wide-Opening Vials
Item Description 150uL Glass Conical with Top Spring

With a nominal O.D. of 6mm, these Thermo Scientific™ National Inserts fit Target LoVial Wide-Opening Vials.

  • Pulled point inserts are an economical choice for noncritical applications
  • Nominal O.D. of 6mm to fit Target wide-opening vials

For use with:

  • Target LoVial Wide-Opening vials
  • Snap-It™ Wide-Opening Snap Cap vials
  • Target DP™ vials
  • Target Wide-Opening vials