Please login to your online account to display your discounted pricing
Thermo Scientific™ National Inserts for Standard-Opening Vials
With a nominal O.D. of 5mm, these Thermo Scientific™ National Inserts fit most standard-opening crimp top vials.
Manufacturer: thermo scientific™  C4012K530
 View more versions of this product
Catalog No. 03-378-359
  • / Pack of 100 Each
Description & Specifications
Specifications
Features Polyspring, Conical, Pulled point, Kimshield™ glass
For Use With Standard-Opening Vials
Type Polyspring, Conical, Pulled point, Kimshield™ glass
Vial Volume (Metric) 200µL
Item Description C4012-K530
With a nominal O.D. of 5mm, these Thermo Scientific™ National Inserts fit most standard-opening crimp top vials.
  • Pulled point inserts are an economical choice for non-critical applications