Please login to your online account to display your discounted pricing
Thermo Scientific™ National Inserts for 15 × 45mm 4mL Vials
Thermo Scientific™ National Vial Inserts offer a variety of options incuding glass conical, Polyspring and pulled point.
Manufacturer: thermo scientific™  C4015641
 View more versions of this product
Catalog No. 03-375-39A
  • / Pack of 100 Each
Description & Specifications
Specifications
Item Description C4015-641
For Use With 15 x 45mm 4mL Vials
Features Polyspring Glass Conical Insert
Type Polyspring Insert, Flange at Insert Lip
Vial Volume (Metric) 500µL
Thermo Scientific™ National Vial Inserts offer a variety of options incuding glass conical, Polyspring and pulled point.
  • Have a nominal O.D. of 6 or 8mm to fit 15 × 45 4mL vials
  • Deactivated inserts are NSC Silanized