Please login to your online account to display your discounted pricing
Thermo Scientific™ National Headspace 20mm Crimp Seals
Thermo Scientific™ National Headspace 20mm Unlined Crimp Seals are compatible with all headspace vials.
$265.83 - $265.83
Specifications
Description One-Piece; Tin-plated steel; Unlined
Closure Color Gold
Closure Material One-Piece; Tin-plated steel; Unlined
Type Crimp Cap Seal
Vial Closure Size 20mm
View More Specs
Products
Catalog Number Mfr. No. Closure Material Price Quantity    
03-376-251
thermo scientific™
C40203M
One-Piece; Tin-plated steel; Unlined Case of 1000 Each for $265.83
Description & Specifications
Specifications
Description One-Piece; Tin-plated steel; Unlined
Closure Color Gold
Closure Material One-Piece; Tin-plated steel; Unlined
Type Crimp Cap Seal
Vial Closure Size 20mm
For Use With All headspace vials
Item Description One-Piece; Tin-plated steel; Unlined
Thermo Scientific™ National Headspace 20mm Unlined Crimp Seals are compatible with all headspace vials.
  • Quick, convenient, and minimize contamination
  • 20mm Crimp seals with prefitted septa for headspace vials
  • Seals with pre-inserted septa are convenient and minimize contamination due to handling
  • Use magnetic seals with CTC/Leap™ Technologies, Gerstel™ and other magnetic transport autosamplers
  • PurePak™ PTFE/Silicone seals are packed in glass jars to maintain purity up to the point of use
  • Molded polyethylene SepCapS is suitable for short-term storage or where repeated access to the vial is required