missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ National Glass Screw-Thread Sample Vials with Open-Top Caps and Septa

Thermo Scientific™ National Glass Screw-Thread Sample Vials with Open-Top Caps and Septa include shrink-wrapped vials and separately packaged caps and septa.

Manufacturer:  Thermo Scientific™ B79904A

 View more versions of this product

Catalog No. 03-391-9E


 

Specifications

Description

Thermo Scientific™ National Glass Screw-Thread Sample Vials with Open-Top Caps and Septa include shrink-wrapped vials and separately packaged caps and septa.

  • Unassembled convenience kits
  • Choice of 33 expansion borosilicate clear glass (Type 1, Class A) or 51A amber glass
  • Eliminate leaching of ions
  • Provide consistent pH for duration of sample storage life
  • With polypropylene open top screw caps and PTFE/silicone septa
  • Convenient packaging for storerooms or laboratory cabinets
Description & Specifications

Specifications

18-400
PTFE/Silicone
21mm
4.375 dr.
Glass
Assembled Vial Kit
16mL
70mm
Amber
Open PP cap w/septa