Please login to your online account to display your discounted pricing

Thermo Scientific™ National Assembled Target™ 10-425 Caps and Septa


Minimize contamination from handling with convenient Thermo Scientific™ National Assembled Target 10-425 Caps and Septa.

Manufacturer: thermo scientific™  C401060AY

 View more versions of this product

 Disclaimers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Catalog No. 03-376-404

  • / Case of 1000 Each
 

Description & Specifications

Specifications

Color White/Red (Septa)
Color Yellow (Cap)
Material Polypropylene (Septa)
Type Open Top
Type Screw Cap with Septum
Product Line National Assembled Target™
Description Yellow; Septa: Red PTFE/White Silicone
Closure Color Yellow
Closure Material Polypropylene
Septum Red PTFE/White Silicone
Item Description C4010-60AY

Minimize contamination from handling with convenient Thermo Scientific™ National Assembled Target 10-425 Caps and Septa.

  • Pre-assembled caps and septa are convenient and minimize contamination from handling
  • Storage cap has a solid top and pre-inserted PTFE disk liner