Please login to your online account to display your discounted pricing
Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits
Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits provide convenience, efficiency and stackable storage options.
Manufacturer: thermo scientific™  C4013491A
 View more versions of this product
Catalog No. 03-378-363
  • / Pack of 100 Each
Description & Specifications
Specifications
Vial Volume (Metric) 2mL
Vial Closure Size 8-425mm
Septa Tan PTFE/White Silicone
Color Clear
Closure Color Black
Type Assembled Vial Kit
Closure Type Phenolic
Item Description C4013-491A
Closure Material Phenolic
Description Clear Glass; Phenolic Black Cap; Tan PTFE/White Silicone; in storage tray
Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits provide convenience, efficiency and stackable storage options.
  • Packaged in convenient vial trays with clear covers or in economical polybags
  • Vial trays are stackable for easy storage
  • All caps are manufactured from virgin polypropylene unless noted
Includes: 
100 vials with pre-attached caps and septa