Please login to your online account to display your discounted pricing
Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits
Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits provide convenience, efficiency and stackable storage options.
Manufacturer: thermo scientific™  C4013019A
 View more versions of this product
Catalog No. 03-375-38B
  • / Pack of 100 Each
Description & Specifications
Specifications
Color Amber
Type Assembled Vial Kit
Closure Color Black
Closure Type Polypropylene
Septa White PTFE
Vial Closure Size 8-425mm
Vial Volume (Metric) 2mL
Closure Material Polypropylene
Item Description C4013-019A
Description Amber Gkass; PP Black Cap; White PTFE septum; in polybag
Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits provide convenience, efficiency and stackable storage options.
  • Packaged in convenient vial trays with clear covers or in economical polybags
  • Vial trays are stackable for easy storage
  • All caps are manufactured from virgin polypropylene unless noted
Includes: 
100 vials with pre-attached caps and septa