Thermo Scientific™ National Assembled Screw Caps with Inserted Septa for 4mL Vials
Thermo Scientific™ National Assembled Screw Caps with Inserted Septa for 4mL Vials are designed to be used with any 13mm septa.
Manufacturer:  Thermo Scientific™ C401575R
 View more versions of this product
Catalog No. 03-376-463
Description
Description
Thermo Scientific™ National Assembled Screw Caps with Inserted Septa for 4mL Vials are designed to be used with any 13mm septa.
Specifications
Specifications
PP, Red; Red PTFE/White Silicone septum
Polypropylene
Open Top Cap
C4015-75R
Red
Red PTFE/White Silicone
4mL Vials
National™
Documents