missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ National Assembled Certified Vial Kits

Thermo Scientific™ National Assembled Certified Vial Kits provide convenience and assurance vials uphold rigorous manufacturing standards.

Manufacturer:  Thermo Scientific™ CERT401091A

 View more versions of this product

Catalog No. 03-377-500


 

Specifications

Description

Thermo Scientific™ National Assembled Certified Vial Kits provide convenience and assurance vials uphold rigorous manufacturing standards.

  • Fully lot tested including HPLC and GC analysis
  • Certificate of Conformance included in every pack
  • Bonded caps to prevent septa push-through
  • Clear Type 1, 33 expansion borosilicate glass or amber Type 2, 51 expansion borosilicate glass
  • Available with or without ID patch
  • Fixed micro vials or inserts for limited-volume sampling
  • Assembled in convenient packs of 100 vials and pre-attached caps and septa
Description & Specifications

Specifications

limited-volume sampling
10mm
Black
Screw Thread
Certified Target DP 2mL Amber Glass Vial Kit, 10mm Screw Thread, Bonded PTFE/Silicone Closure
Amber
Assembled Vial Kit
2mL
Bonded PTFE/Silicone
2mL Amber Vial Kit, 10mm Screw Thread
Bonded PTFE/Silicone Septum